Assay Details
| Gene Symbol: Dusp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105734.1 | NP_001099204.1 | 3 - 4 | 1339 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of adaptive immune response
protein dephosphorylation
response to organic substance
response to lipopolysaccharide
negative regulation of stress-activated MAPK cascade
peptidyl-tyrosine dephosphorylation
negative regulation of JUN kinase activity
negative regulation of protein kinase activity by regulation of protein phosphorylation
regulation of innate immune response
negative regulation of JNK cascade
oligodendrocyte differentiation
negative regulation of oligodendrocyte differentiation
negative regulation of respiratory burst involved in inflammatory response
regulation of brown fat cell differentiation
molecular_function
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MAP kinase phosphatase activity
inactivation of MAPK activity
regulation of adaptive immune response
protein dephosphorylation
response to organic substance
response to lipopolysaccharide
negative regulation of stress-activated MAPK cascade
peptidyl-tyrosine dephosphorylation
negative regulation of JUN kinase activity
negative regulation of protein kinase activity by regulation of protein phosphorylation
regulation of innate immune response
negative regulation of JNK cascade
oligodendrocyte differentiation
negative regulation of oligodendrocyte differentiation
negative regulation of respiratory burst involved in inflammatory response
regulation of brown fat cell differentiation
molecular_function
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MAP kinase phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105216 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026152 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026152 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026152 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P264892 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10766465 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766466 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766464 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766467 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10766463 | - | - |
| Rat Genome 230 2.0 Array | 1394028_at | - | - |
| Rat Genome 230 2.0 Array | 1394028_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394028_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394028_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300007118.rn.1 | TC1300000985.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007117.rn.1 | TC1300000985.rn.1 | RTA TC |
| Rat Expression Set 230 | 1394028_PM_at | - | - |
| Rat Expression Set 230 | 1394028_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21407389 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365349 | - | - |

Dusp10
Genomic Map