Assay Details
| Gene Symbol: Cst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019258.1 | NP_062131.1 | 1 - 2 | 112 | 69 | |
| GenBank mRNA | AF090692.1 | - | 1 - 2 | 112 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF090692.1, NM_019258.1 transcripts.
Set Membership: |
|

Cst8
Genomic Map