Assay Details
| Gene Symbol: Rbbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019219.2 | NP_062092.2 | 3 - 4 | 267 | 76 | |
| GenBank mRNA | AF025819.2 | - | 3 - 4 | 267 | 76 | |
| BC101915.1 | - | 3 - 4 | 311 | 7935765 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbbp9
Genomic Map