Assay Details
| Gene Symbol: Snrpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108592.2 | NP_001102062.1 | 5 - 6 | 484 | 150 | |
| XM_006235116.2 | XP_006235178.1 | 5 - 6 | 645 | 150 | ||
| GenBank mRNA | BC168760.1 | - | 5 - 6 | 484 | 6888886 | 150 |
| FQ213679.1 | - | 5 - 6 | 516 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snrpb2
Genomic Map