Assay Details
Gene Symbol: Cst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012837.1 | NP_036969.1 | 1 - 2 | 298 | 121 | |
GenBank mRNA | BC087591.1 | - | 1 - 2 | 238 | 7307694 | 121 |
CB314369.1 | - | 1 - 2 | 70 | 121 | ||
CB608544.1 | - | 1 - 2 | 281 | 121 | ||
FQ210306.1 | - | 1 - 2 | 251 | 121 | ||
FQ211221.1 | - | 1 - 2 | 251 | 121 | ||
FQ211526.1 | - | 1 - 2 | 251 | 121 | ||
FQ211840.1 | - | 1 - 2 | 251 | 121 | ||
FQ211941.1 | - | 1 - 2 | 251 | 121 | ||
FQ212057.1 | - | 1 - 2 | 280 | 121 | ||
FQ212112.1 | - | 1 - 2 | 251 | 121 | ||
FQ212277.1 | - | 1 - 2 | 251 | 121 | ||
FQ212367.1 | - | 1 - 2 | 251 | 121 | ||
FQ214337.1 | - | 1 - 2 | 121 | |||
FQ218053.1 | - | 1 - 2 | 282 | 121 | ||
FQ221232.1 | - | 1 - 2 | 251 | 121 | ||
FQ221782.1 | - | 1 - 2 | 251 | 121 | ||
FQ222268.1 | - | 1 - 2 | 251 | 121 | ||
FQ222820.1 | - | 1 - 2 | 251 | 121 | ||
FQ223224.1 | - | 1 - 2 | 251 | 121 | ||
FQ224103.1 | - | 1 - 2 | 251 | 121 | ||
FQ228332.1 | - | 1 - 2 | 248 | 121 | ||
FQ228399.1 | - | 1 - 2 | 251 | 121 | ||
FQ228646.1 | - | 1 - 2 | 251 | 121 | ||
FQ234324.1 | - | 1 - 2 | 282 | 121 | ||
X16957.1 | - | 1 - 2 | 192 | 121 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
