Assay Details
| Gene Symbol: Napb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191966.1 | NP_001178895.1 | 8 - 9 | 667 | 91 | |
| XM_008762275.1 | XP_008760497.1 | 6 - 7 | 522 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Napb
Genomic Map