Assay Details
| Gene Symbol: Csrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191855.1 | NP_001178784.1 | 5 - 6 | 1356 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csrp2bp| Gene Symbol: Csrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191855.1 | NP_001178784.1 | 5 - 6 | 1356 | 64 | |
Set Membership: |
|