Assay Details
Gene Symbol: Cst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153734.1 | NP_714956.1 | 1 - 2 | 242 | 116 | |
GenBank mRNA | AF440736.1 | - | 1 - 2 | 242 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
