Assay Details
| Gene Symbol: Bfsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031555.1 | NP_113743.1 | 3 - 4 | 532 | 63 | |
| XM_006235117.2 | XP_006235179.1 | 3 - 4 | 193 | 63 | ||
| GenBank mRNA | AB003104.1 | - | 3 - 4 | 533 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bfsp1
Genomic Map