Assay Details
| Gene Symbol: Gzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107788.1 | NP_001101258.1 | 4 - 5 | 1887 | 82 | |
| XM_006235153.2 | XP_006235215.1 | 4 - 5 | 1754 | 82 | ||
| XM_006235154.2 | XP_006235216.1 | 4 - 5 | 2866 | 82 | ||
| XM_008762274.1 | XP_008760496.1 | 5 - 6 | 2045 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gzf1
Genomic Map