Assay Details
| Gene Symbol: Rin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107786.1 | NP_001101256.1 | 7 - 8 | 803 | 68 | |
| XM_006235142.2 | XP_006235204.1 | 7 - 8 | 2378 | 68 | ||
| XM_006235143.2 | XP_006235205.1 | 6 - 7 | 1660 | 68 | ||
| XM_006235146.2 | XP_006235208.1 | 6 - 7 | 1330 | 68 | ||
| XM_006235147.2 | XP_006235209.1 | 6 - 7 | 1433 | 68 | ||
| XM_006235148.2 | XP_006235210.1 | 5 - 6 | 984 | 68 | ||
| XM_006235149.2 | XP_006235211.1 | 6 - 7 | 1747 | 68 | ||
| XM_006235150.2 | XP_006235212.1 | 5 - 6 | 998 | 68 | ||
| XM_006235151.2 | XP_006235213.1 | 4 - 5 | 488 | 68 | ||
| XM_008762270.1 | XP_008760492.1 | 8 - 9 | 1821 | 68 | ||
| XM_008762271.1 | XP_008760493.1 | 6 - 7 | 1216 | 68 | ||
| XM_008762272.1 | XP_008760494.1 | 4 - 5 | 1238 | 68 | ||
| XM_008762273.1 | XP_008760495.1 | 4 - 5 | 2259 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rin2
Genomic Map