Assay Details
| Gene Symbol: Mtg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013924.1 | NP_001013946.1 | 4 - 5 | 558 | 84 | |
| XM_006235817.2 | XP_006235879.1 | 4 - 5 | 984 | 84 | ||
| XM_006235818.2 | XP_006235880.1 | 4 - 5 | 604 | 84 | ||
| XM_006235820.2 | XP_006235882.1 | 4 - 5 | 612 | 84 | ||
| XM_008762485.1 | XP_008760707.1 | 3 - 4 | 2118 | 84 | ||
| XM_008762486.1 | XP_008760708.1 | 4 - 5 | 988 | 84 | ||
| XM_008762487.1 | XP_008760709.1 | 5 - 6 | 705 | 84 | ||
| XM_008762488.1 | XP_008760710.1 | 5 - 6 | 728 | 84 | ||
| XM_008762489.1 | XP_008760711.1 | 4 - 5 | 627 | 84 | ||
| XM_008762490.1 | XP_008760712.1 | 5 - 6 | 684 | 84 | ||
| XM_008762491.1 | XP_008760713.1 | 4 - 5 | 608 | 84 | ||
| XM_008762492.1 | XP_008760714.1 | 3 - 4 | 2118 | 84 | ||
| XM_008762493.1 | XP_008760715.1 | 4 - 5 | 635 | 84 | ||
| XM_008762494.1 | XP_008760716.1 | 3 - 4 | 2118 | 84 | ||
| XM_008762495.1 | XP_008760717.1 | 3 - 4 | 2118 | 84 | ||
| GenBank mRNA | BC083707.1 | - | 4 - 5 | 558 | 7190542 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Mtg2
Genomic Map