Assay Details
Gene Symbol: Lkaaear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106551.1 | NP_001100021.1 | 2 - 3 | 433 | 59 | |
XM_006235763.2 | XP_006235825.1 | 2 - 3 | 543 | 59 | ||
XM_006235764.2 | XP_006235826.1 | 2 - 3 | 482 | 59 | ||
XM_008762501.1 | XP_008760723.1 | 2 - 3 | 411 | 59 | ||
XM_008762502.1 | XP_008760724.1 | 1 - 2 | 405 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
