Assay Details
| Gene Symbol: Piga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108816.2 | NP_001102286.1 | 2 - 3 | 154 | 95 | |
| XM_006256870.2 | XP_006256932.1 | 3 - 4 | 2404 | 95 | ||
| XM_006256873.2 | XP_006256935.1 | 3 - 4 | 982 | 95 | ||
| XM_006256875.2 | XP_006256937.1 | 3 - 4 | 2524 | 95 | ||
| XM_006256876.1 | XP_006256938.1 | 3 - 4 | 1038 | 95 | ||
| XM_008773175.1 | XP_008771397.1 | 2 - 3 | 3702 | 95 | ||
| XM_008773176.1 | XP_008771398.1 | 3 - 4 | 3385 | 95 | ||
| XM_008773177.1 | XP_008771399.1 | 3 - 4 | 2973 | 95 | ||
| XM_008773178.1 | XP_008771400.1 | 3 - 4 | 3441 | 95 | ||
| XM_008773179.1 | XP_008771401.1 | 3 - 4 | 2449 | 95 | ||
| XM_008773180.1 | XP_008771402.1 | 3 - 4 | 2505 | 95 | ||
| XM_008773181.1 | XP_008771403.1 | 3 - 4 | 1506 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Piga
Genomic Map