Assay Details
| Gene Symbol: Trappc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024965.1 | NP_001020136.1 | 2 - 3 | 89 | 89 | |
| XM_006256857.1 | XP_006256919.1 | 2 - 3 | 179 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001024965.1, XM_006256857.1 transcripts.
Set Membership: |
|

Trappc2
Genomic Map