Assay Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001069267.4 | XP_001069267.2 | 3 - 4 | 510 | 96 | |
XM_006238551.2 | XP_006238613.1 | 3 - 4 | 512 | 96 | ||
XM_008763922.1 | XP_008762144.1 | 3 - 4 | 499 | 96 | ||
XM_008763923.1 | XP_008762145.1 | 3 - 4 | 500 | 96 | ||
XM_008763924.1 | XP_008762146.1 | 3 - 4 | 498 | 96 | ||
XM_008763925.1 | XP_008762147.1 | 3 - 4 | 486 | 96 | ||
XM_008763926.1 | XP_008762148.1 | 3 - 4 | 490 | 96 | ||
XM_008763927.1 | XP_008762149.1 | 3 - 4 | 494 | 96 | ||
XM_008763929.1 | XP_008762151.1 | 3 - 4 | 511 | 96 | ||
XM_008763930.1 | XP_008762152.1 | 3 - 4 | 610 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
