Assay Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001069267.4 | XP_001069267.2 | 6 - 7 | 909 | 115 | |
XM_008763922.1 | XP_008762144.1 | 6 - 7 | 898 | 115 | ||
XM_008763923.1 | XP_008762145.1 | 6 - 7 | 899 | 115 | ||
XM_008763924.1 | XP_008762146.1 | 6 - 7 | 897 | 115 | ||
XM_008763925.1 | XP_008762147.1 | 6 - 7 | 885 | 115 | ||
XM_008763926.1 | XP_008762148.1 | 6 - 7 | 889 | 115 | ||
XM_008763927.1 | XP_008762149.1 | 6 - 7 | 893 | 115 | ||
XM_008763928.1 | XP_008762150.1 | 5 - 6 | 832 | 115 | ||
XM_008763929.1 | XP_008762151.1 | 5 - 6 | 805 | 115 | ||
XM_008763930.1 | XP_008762152.1 | 6 - 7 | 1009 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
