Assay Details
Gene Symbol: Gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034913.1 | NP_001030085.1 | 10 - 11 | 1628 | 90 | |
GenBank mRNA | BC105913.1 | - | 10 - 11 | 1628 | 7385857 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55157 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012919 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012919 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012919 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48816 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93807 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10871251 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934578 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774436 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871254 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742107 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871256 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742106 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871258 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774437 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871252 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871255 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871257 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742105 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871253 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833672 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10871250 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10774435 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10819130 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10742104 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10833668 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10934577 | - | - |
Rat Genome 230 2.0 Array | 1367542_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376776_at | - | - |
Rat Genome 230 2.0 Array | 1376776_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367542_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500008752.rn.1 | TC0500001266.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500008754.rn.1 | TC0500001266.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500008753.rn.1 | TC0500001266.rn.1 | RTA TC |
Rat Expression Set 230 | 1367542_PM_at | - | - |
Rat Expression Set 230 | 1367542_at | - | - |
Rat Expression Set 230 | 1367542_PM_at | - | - |
Rat Expression Set 230 | 1367542_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742104 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871250 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10833668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10774435 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819130 | - | - |
Rat Genome U34 Set | rc_AA900343_f_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21022287 | - | - |
ABI Rat Genome Survey Microarray | 22406367 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358520 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650756 | - | - |
