Assay Details
| Gene Symbol: Txndc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100840.1 | NP_001094310.1 | 2 - 3 | 296 | 87 | |
| GenBank mRNA | BC100255.1 | - | 2 - 3 | 238 | 87 | |
| BC158737.1 | - | 2 - 3 | 242 | 87 | ||
| CV117277.1 | - | 2 - 3 | 533 | 87 | ||
| FQ218354.1 | - | 2 - 3 | 193 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txndc12
Genomic Map