Assay Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138827.1 | NP_620182.1 | 8 - 9 | 1281 | 73 | |
| GenBank mRNA | BC061873.1 | - | 8 - 9 | 1209 | 5598829 | 73 |
| M13979.1 | - | 8 - 9 | 1281 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
response to osmotic stress
glucose transport
proton transport
cellular response to glucose starvation
xenobiotic transport
transmembrane transport
dehydroascorbic acid transport
sugar:proton symporter activity
glucose transmembrane transporter activity
protein binding
kinase binding
dehydroascorbic acid transporter activity
identical protein binding
xenobiotic transporter activity
protein self-association
D-glucose transmembrane transporter activity
protein complex assembly
response to osmotic stress
glucose transport
proton transport
cellular response to glucose starvation
xenobiotic transport
transmembrane transport
dehydroascorbic acid transport
sugar:proton symporter activity
glucose transmembrane transporter activity
protein binding
kinase binding
dehydroascorbic acid transporter activity
identical protein binding
xenobiotic transporter activity
protein self-association
D-glucose transmembrane transporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P684538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39675 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P684538 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P684538 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P684538 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P684538 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700526341 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702140296 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15711 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64377 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5730 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702140296 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700526341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10871531 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871527 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871528 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871525 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10871521 | - | - |
| Rat Genome 230 2.0 Array | 1370848_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370848_at | - | - |
| Affymetrix HT Rat Focus Array | 1370848_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370848_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500009350.rn.1 | TC0500001342.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370848_at | - | - |
| Rat Expression Set 230 | 1370848_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871521 | - | - |
| Rat Genome U34 Set | M13979_at | - | - |
| Rat Genome U34 Set | S68135_s_at | - | - |
| Rat Genome U34 Set | rc_AI008668_s_at | - | - |
| Rat Genome U34 Set | AA944077_s_at | - | - |
| Rat Genome U34 Set | S68135_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20706716 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355214 | - | - |
| RatRef-12 Expression BeadChip | ILMN_56823 | - | - |

Slc2a1
Genomic Map