Assay Details
| Gene Symbol: Kti12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044288.1 | NP_001037753.1 | 1 | 266 | 67 | |
| GenBank mRNA | BC088196.1 | - | 1 | 266 | 7374597 | 67 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kti12
Genomic Map