Assay Details
| Gene Symbol: Cyp4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238712.2 | XP_006238774.1 | 12 - 13 | 1585 | 130 | |
| GenBank mRNA | X07259.1 | - | 12 - 13 | 1543 | 130 | |
More Information
Set Membership: |
|

Cyp4a1
Genomic Map