Assay Details
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729532.4 | XP_002729578.2 | 4 - 5 | 544 | 97 | |
XM_006238747.2 | XP_006238809.1 | 4 - 5 | 546 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
