Assay Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729532.4 | XP_002729578.2 | 5 - 6 | 589 | 105 | |
| XM_006238747.2 | XP_006238809.1 | 5 - 6 | 591 | 105 | ||
More Information
Set Membership: |
|

Hyi
Genomic Map