Assay Details
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729532.4 | XP_002729578.2 | 6 - 7 | 665 | 62 | |
XM_006238748.2 | XP_006238810.1 | 5 - 6 | 617 | 62 | ||
GenBank mRNA | FQ211344.1 | - | 4 - 5 | 519 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
