Assay Details
| Gene Symbol: Toe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106680.1 | NP_001100150.1 | 6 - 7 | 886 | 74 | |
| XM_006238651.1 | XP_006238713.1 | 6 - 7 | 881 | 74 | ||
| XM_006238652.2 | XP_006238714.1 | 5 - 6 | 1211 | 74 | ||
| XM_006238653.2 | XP_006238715.1 | 5 - 6 | 1133 | 74 | ||
| XM_006238654.2 | XP_006238716.1 | 5 - 6 | 1234 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Toe1
Genomic Map