Assay Details
Gene Symbol: Ybx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031563.3 | NP_113751.3 | 1 - 2 | 280 | 113 | |
XM_008764058.1 | XP_008762280.1 | 1 - 2 | 320 | 113 | ||
XM_008764059.1 | XP_008762281.1 | 1 - 2 | 520 | 113 | ||
GenBank mRNA | AB046928.2 | - | 1 - 2 | 439 | 113 | |
BC072486.1 | - | 1 - 2 | 280 | 7105037 | 113 | |
BC098672.1 | - | 1 - 2 | 280 | 7124218 | 113 | |
BC103637.1 | - | 1 - 2 | 274 | 7107271 | 113 | |
D13309.1 | - | 1 - 2 | 250 | 113 | ||
M57299.1 | - | 1 - 2 | 256 | 113 | ||
M69138.1 | - | 1 - 2 | 416 | 113 |
Gene Symbol: Ybx1-ps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038098.1 | 1 | 291 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
