Assay Details
Gene Symbol: Rfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053547.1 | NP_445999.1 | 2146 | 63 | ||
XM_006250972.2 | XP_006251034.1 | 2208 | 63 | |||
XM_008770161.1 | XP_008768383.1 | 2211 | 63 | |||
XM_008770162.1 | XP_008768384.1 | 2208 | 63 | |||
GenBank mRNA | AF030050.1 | - | 14 - 15 | 2066 | 63 | |
AF059678.1 | - | 2152 | 63 | |||
CV110819.1 | - | 3 - 4 | 390 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
