Assay Details
| Gene Symbol: Glce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_343404.6 | XP_343405.2 | 2 - 3 | 297 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_343404.6 transcripts.
Set Membership: |
|

Glce
Genomic Map