Assay Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025123.3 | NP_001020294.3 | 6 - 7 | 1300 | 111 | |
| GenBank mRNA | BC093594.1 | - | 6 - 7 | 1300 | 7314531 | 111 |
| BC100617.1 | - | 6 - 7 | 1299 | 7934552 | 111 | |
| FQ211817.1 | - | 5 - 6 | 1004 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kdelc2
Genomic Map