Assay Details
| Gene Symbol: Hspa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024351.2 | NP_077327.1 | 5 - 6 | 1202 | 154 | |
| GenBank mRNA | BC061547.1 | - | 5 - 6 | 1209 | 6921606 | 154 |
| BC098914.1 | - | 5 - 6 | 1154 | 7104230 | 154 | |
| FQ213866.1 | - | 1 - 2 | 218 | 154 | ||
| FQ231904.1 | - | 1 - 2 | 394 | 154 | ||
| FQ232261.1 | - | 1 - 2 | 137 | 154 | ||
| FQ235115.1 | - | 1 - 2 | 551 | 154 | ||
| M11942.1 | - | 5 - 6 | 1173 | 154 | ||
| Gene Symbol: LOC689955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764620.1 | XP_008762842.1 | 1 | 1204 | 154 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hspa8
Genomic Map