Assay Details
Gene Symbol: LOC689599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750754.3 | XP_003750802.1 | 2 - 3 | 354 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC689599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750754.3 | XP_003750802.1 | 2 - 3 | 354 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() |