Assay Details
Gene Symbol: Prrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191888.1 | NP_001178817.1 | 4 - 5 | 366 | 115 | |
XM_006256963.2 | XP_006257025.1 | 5 - 6 | 530 | 115 | ||
XM_006256965.2 | XP_006257027.1 | 3 - 4 | 360 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
