Assay Details
| Gene Symbol: Rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104643.1 | NP_001098113.1 | 3 - 4 | 187 | 130 | |
| GenBank mRNA | EU074986.1 | - | 3 - 4 | 187 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rs1
Genomic Map