Assay Details
| Gene Symbol: Dydc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252813.2 | XP_006252875.1 | 1 - 2 | 244 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006252813.2 transcripts.
Set Membership: |
|

Dydc2
Genomic Map