Assay Details
| Gene Symbol: Mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106072.2 | NP_001099542.1 | 2 - 3 | 415 | 80 | |
| XM_008771087.1 | XP_008769309.1 | 2 - 3 | 420 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpv17l2
Genomic Map