Assay Details
| Gene Symbol: Rpl18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212510.1 | NP_997675.1 | 3 - 4 | 374 | 91 | |
| GenBank mRNA | BC058498.1 | - | 3 - 4 | 374 | 6919102 | 91 |
| FQ209874.1 | - | 2 - 3 | 215 | 91 | ||
| FQ209881.1 | - | 3 - 4 | 368 | 91 | ||
| FQ209956.1 | - | 3 - 4 | 374 | 91 | ||
| FQ216877.1 | - | 3 - 4 | 367 | 91 | ||
| FQ221165.1 | - | 3 - 4 | 369 | 91 | ||
| FQ221243.1 | - | 3 - 4 | 368 | 91 | ||
| FQ221413.1 | - | 3 - 4 | 368 | 91 | ||
| FQ221831.1 | - | 3 - 4 | 375 | 91 | ||
| FQ222162.1 | - | 3 - 4 | 368 | 91 | ||
| FQ222289.1 | - | 3 - 4 | 368 | 91 | ||
| FQ222474.1 | - | 3 - 4 | 372 | 91 | ||
| FQ222662.1 | - | 3 - 4 | 368 | 91 | ||
| FQ223818.1 | - | 3 - 4 | 369 | 91 | ||
| FQ228440.1 | - | 3 - 4 | 368 | 91 | ||
| X14181.1 | - | 3 - 4 | 368 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl18a
Genomic Map