Assay Details
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053998.2 | NP_446450.2 | 2 - 3 | 191 | 146 | |
GenBank mRNA | BC071176.1 | - | 2 - 3 | 184 | 146 | |
BC105863.1 | - | 2 - 3 | 191 | 7457017 | 146 | |
JN019799.1 | - | 2 - 3 | 186 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
