Assay Details
| Gene Symbol: Use1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169100.1 | NP_001162571.1 | 3 - 4 | 268 | 71 | |
| XM_006252855.2 | XP_006252917.1 | 3 - 4 | 1188 | 71 | ||
| GenBank mRNA | BC158812.1 | - | 3 - 4 | 244 | 71 | |
| FQ217580.1 | - | 3 - 4 | 269 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Use1
Genomic Map