Assay Details
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106070.1 | NP_001099540.1 | 2 - 3 | 263 | 101 | |
XM_008771085.1 | XP_008769307.1 | 2 - 3 | 246 | 101 | ||
XM_008771086.1 | XP_008769308.1 | 2 - 3 | 246 | 101 | ||
GenBank mRNA | BC169058.1 | - | 2 - 3 | 235 | 7932017 | 101 |
FQ211658.1 | - | 2 - 3 | 251 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008771085.1, XM_008771086.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
