Assay Details
| Gene Symbol: Pgpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201988.2 | NP_973717.1 | 2 - 3 | 103 | 80 | |
| GenBank mRNA | AB098134.1 | - | 2 - 4 | 80 | ||
| BC098645.1 | - | 2 - 3 | 103 | 7108516 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgpep1
Genomic Map