Assay Details
Gene Symbol: Abhd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107301.1 | NP_001100771.1 | 2 - 3 | 927 | 60 | |
GenBank mRNA | BC168736.1 | - | 2 - 3 | 868 | 6891332 | 60 |
BC168761.1 | - | 2 - 3 | 868 | 6889450 | 60 | |
BC168765.1 | - | 2 - 3 | 868 | 6889859 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
