Assay Details
| Gene Symbol: LOC100911483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277222.1 | NP_001264151.1 | 3 - 4 | 242 | 114 | |
| GenBank mRNA | FM065403.1 | - | 3 - 4 | 238 | 114 | |
| FQ211515.1 | - | 3 - 4 | 310 | 114 | ||
| FQ221417.1 | - | 3 - 4 | 247 | 114 | ||
| FQ223566.1 | - | 3 - 4 | 253 | 114 | ||
| FQ223934.1 | - | 3 - 4 | 267 | 114 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911483
Genomic Map