Assay Details
| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106064.1 | NP_001099534.1 | 3 - 4 | 524 | 81 | |
| XM_006252796.2 | XP_006252858.1 | 3 - 4 | 458 | 81 | ||
| XM_006252797.2 | XP_006252859.1 | 3 - 4 | 458 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cherp
Genomic Map