Assay Details
| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145085.2 | NP_659553.1 | 1 - 2 | 154 | 126 | |
| GenBank mRNA | AF332142.1 | - | 144 | 126 | ||
| BC078719.1 | - | 1 - 2 | 154 | 7127920 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyox1
Genomic Map