Assay Details
Gene Symbol: Fam19a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134700.1 | NP_001128172.1 | 4 - 5 | 622 | 71 | |
GenBank mRNA | BC160856.1 | - | 4 - 5 | 622 | 8364977 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P855071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178070 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P855071 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P855071 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P855071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94892 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99511 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30233 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3668 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11004 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29772 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98063 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22626 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10864329 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864328 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864330 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10864327 | - | - |
Rat Genome 230 2.0 Array | 1378557_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378557_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400024123.rn.1 | TC0400003410.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024124.rn.1 | TC0400003410.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400024125.rn.1 | TC0400003410.rn.1 | RTA TC |
Rat Expression Set 230 | 1378557_PM_at | - | - |
Rat Expression Set 230 | 1378557_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864327 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20814180 | - | - |
ABI Rat Genome Survey Microarray | 21516852 | - | - |
