Assay Details
Gene Symbol: Trh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013046.3 | NP_037178.1 | 1 - 2 | 123 | 55 | |
GenBank mRNA | BC161830.1 | - | 1 - 2 | 70 | 9091055 | 55 |
FQ100942.1 | - | 1 - 2 | 123 | 55 | ||
FQ221570.1 | - | 1 - 2 | 99 | 55 | ||
FQ221573.1 | - | 1 - 2 | 99 | 55 | ||
FQ222015.1 | - | 1 - 2 | 99 | 55 | ||
FQ222264.1 | - | 1 - 2 | 97 | 55 | ||
FQ222377.1 | - | 1 - 2 | 98 | 55 | ||
FQ222571.1 | - | 1 - 2 | 97 | 55 | ||
FQ222984.1 | - | 1 - 2 | 97 | 55 | ||
FQ223091.1 | - | 1 - 2 | 99 | 55 | ||
FQ228499.1 | - | 1 - 2 | 99 | 55 | ||
FQ228758.1 | - | 1 - 2 | 99 | 55 | ||
FQ228762.1 | - | 1 - 2 | 97 | 55 | ||
FQ229082.1 | - | 1 - 2 | 99 | 55 | ||
FQ229292.1 | - | 1 - 2 | 97 | 55 | ||
M12138.1 | - | 1 - 2 | 90 | 55 | ||
M36317.1 | - | 1 - 2 | 90 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC161830.1, M12138.1, M36317.1, NM_013046.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
