Assay Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014188.1 | NP_001014210.1 | 1 - 2 | 84 | 101 | |
| GenBank mRNA | BC079119.1 | - | 1 - 2 | 84 | 7126525 | 101 |
| FQ218201.1 | - | 1 - 2 | 135 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079119.1, NM_001014188.1 transcripts.
Set Membership: |
|

Isy1
Genomic Map