Assay Details
Gene Symbol: Rab7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023950.3 | NP_076440.1 | 1 - 2 | 163 | 62 | |
GenBank mRNA | AF286535.1 | - | 1 - 2 | 159 | 62 | |
BC072470.1 | - | 1 - 2 | 163 | 7098657 | 62 | |
X12535.1 | - | 1 - 2 | 36 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF286535.1, BC072470.1, NM_023950.3, X12535.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
