Assay Details
| Gene Symbol: Abcg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106816.1 | NP_001100286.1 | 2 - 3 | 564 | 89 | |
| XM_006242915.2 | XP_006242977.1 | 2 - 3 | 6296 | 89 | ||
| XM_006242916.2 | XP_006242978.1 | 2 - 3 | 654 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg4
Genomic Map